Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium phosphate, dibasic, trihydrate, extra pure

Catalog No. p-4813619
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447732500 250 g Plastic bottle
AC447730010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447732500 Supplier Thermo Scientific Chemicals Supplier No. 447732500
Only null left
Add to Cart

CAS: 16788-57-1 | H7K2O7P | 228.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
IUPAC Name dipotassium;hydrogen phosphate;trihydrate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O

Specifications

Quantity 250 g
Linear Formula K2HPO4·3H2O
Solubility Information Solubility in water: freely soluble
Formula Weight 228.23
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Potassium phosphate, dibasic, trihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.