Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Potassium Phosphate, Dibasic, Trihydrate, Molecular Biology Grade, Calbiochem™,

Catalog No. 5295671KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-956-71KG 1 kg
52-956-7250GM 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-956-71KG Supplier MilliporeSigma™ Supplier No. 5295671KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16788-57-1 | Formula: H7K2O7P | Weight: 228.22 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
IUPAC Name dipotassium trihydrate hydrogen phosphate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O

Specifications

Physical Form Solid
Quantity 1 kg
Assay Percent Range ≥99%
Solubility Information Easily soluble in cold water
Formula Weight 228.2
Percent Purity ≥98%
Grade by titration
Chemical Name or Material Potassium Phosphate, Dibasic, Trihydrate
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.