Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Dibasic Trihydrate, MP Biomedicals™

Catalog No. MP021914325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021914325 500 g
MP021914321 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021914325 Supplier MP Biomedicals, Inc Supplier No. 02191432.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Widely used in molecular biology, biochemistry and chromatography. Potassium Phosphate Dibasic Trihydrate, MP Biomedicals is a reagent with high buffering capacity. It is typically used as a component in microorganism culture media and in phosphate buffer

Potassium phosphate occurs in several forms: monobasic, dibasic and tribasic. Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.

  • It is typically used as a component for a wide variety of media used in the culture of microorganisms
  • It is a component in phosphate buffered saline (PBS)
  • In addition to helping maintain pH, it supplies essential phosphate
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
IUPAC Name dipotassium trihydrate hydrogen phosphate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Solubility Information Soluble in water (100mg/mL); Insoluble to slightly soluble in ethanol; insoluble in alcohol.
Formula Weight 228.18g/mol
Percent Purity 99.8%
Grade Not Available
Chemical Name or Material Potassium Phosphate
Recommended Storage Store at Room Temperature(15–30 °C), desiccate.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.