Learn More
Strem, An Ascensus Company CAS# 7778-53-2. 1kg. Potassium phosphate, anhydrous, min. 97%. MFCD00036295
SDP
Catalog No. 5090110648
Encompass
CAS# 7778-53-2. 1kg. Potassium phosphate, anhydrous, min. 97%. MFCD00036295. Molecular Weight: 212.28. Molecular Formula: K3PO4. Color/form: coarse white solid Strem# 19-3800. http://www.strem.com/catalog/v/19-3800/
Catalog No. 50-901-10648 Supplier Strem, An Ascensus Company Supplier No. 1938001000G
CAS# 7778-53-2. 1kg. Potassium phosphate, anhydrous, min. 97%. MFCD00036295. Molecular Weight: 212.28. Molecular Formula: K3PO4. Color/form: coarse white solid Strem# 19-3800. http://www.strem.com/catalog/v/19-3800/