Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium phosphate monoba25KG
Catalog No. 502902559
Encompass
Potassium phosphate monobasic (monopotassium orthophosphate) is a water soluble inorganic salt. Its standard enthalpy of formation has been reported to be -376.1kcalmol-1. It acts a starting material in the synthesis of potassium metaphosphate.
Catalog No. 50-290-2559 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P06622.5KG
Potassium phosphate monobasic (monopotassium orthophosphate) is a water soluble inorganic salt. Its standard enthalpy of formation has been reported to be -376.1kcalmol-1. It acts a starting material in the synthesis of potassium metaphosphate.