Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium phosphate monobasic

Catalog No. 502907860
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-7860 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-290-7860 Supplier Sigma Aldrich Supplier No. 229806250G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium phosphate monobasic
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 252.6°C
CAS 7778-77-0
Quantity 250 g
Molecular Formula H2KO4P
Linear Formula KH2PO4
MDL Number MFCD00011401
Synonym Monopotassium phosphate; Potassium dihydrogen phosphate; prim.-Potassium phosphate
Molecular Weight (g/mol) 136.09
Percent Purity 99.99% trace metals basis
Density 2.338 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.