Learn More
VWR INTERNATIONAL LLC Potassium dihydrogen phosphate, 7778-77-0, MFCD00011401, 500G
Catalog No. NC0560013
Encompass
Formula: KH₂PO₄, MW: 136.09 g/mol, Boiling Pt: ∼100 °C (1013 hPa), Melting Pt: 253 °C, Storage Temperature: Ambient, Purity: 99%, Synonyms: Potassium dihydrogen orthophosphate , Potassium phosphate monobasic , Monobasic Potassium Phosphate , Potassium orthophosphate monobasic , Potassium dihydrogenorthophosphate.
Catalog No. NC0560013 Supplier VWR INTERNATIONAL LLC Supplier No. 97062350
Formula: KH₂PO₄, MW: 136.09 g/mol, Boiling Pt: ∼100 °C (1013 hPa), Melting Pt: 253 °C, Storage Temperature: Ambient, Purity: 99%, Synonyms: Potassium dihydrogen orthophosphate , Potassium phosphate monobasic , Monobasic Potassium Phosphate , Potassium orthophosphate monobasic , Potassium dihydrogenorthophosphate.