Learn More
Alkali Scientific Potassium Phosphate, Monobasic, ACS Grade, 2.5 Kg
SDP
Catalog No. 5000661303
Encompass_Preferred
Potassium Phosphate Monobasic, ACS Grade, in a 2.5 kg container is a high-purity reagent commonly used in preparing phosphate-buffered solutions in scientific research and industrial laboratories. This salt is essential in creating buffers that maintain stable pH conditions for biochemical assays, nucleic acid research, and microbial cultivation. Potassium Phosphate Monobasic is widely used in enzyme catalysis, protein electrophoresis, and molecular biology experiments. The 2.5 kg volume is perfect for laboratories that require a larger quantity of reagent for frequent buffer preparation, ensuring efficient and consistent pH control in multiple experimental processes.
Catalog No. 50-006-61303 Supplier Alkali Scientific Supplier No. POA006CP
Potassium Phosphate Monobasic, ACS Grade, in a 2.5 kg container is a high-purity reagent commonly used in preparing phosphate-buffered solutions in scientific research and industrial laboratories. This salt is essential in creating buffers that maintain stable pH conditions for biochemical assays, nucleic acid research, and microbial cultivation. Potassium Phosphate Monobasic is widely used in enzyme catalysis, protein electrophoresis, and molecular biology experiments. The 2.5 kg volume is perfect for laboratories that require a larger quantity of reagent for frequent buffer preparation, ensuring efficient and consistent pH control in multiple experimental processes.