Learn More
Alkali Scientific Potassium Phosphate, Monobasic, ACS Grade, 250 G
SDP
Catalog No. 5000661302
Encompass_Preferred
Potassium Phosphate Monobasic, ACS Grade, in a 250g volume is a versatile laboratory reagent used to prepare buffer solutions for a wide range of biochemical, microbiological, and chemical applications. It is especially useful in preparing phosphate buffers and phosphate-buffered saline (PBS) for enzymatic reactions, protein assays, and nucleic acid extractions. Potassium Phosphate Monobasic helps maintain the desired pH in various laboratory processes, ensuring consistency and reliability in experimental results. This smaller volume of 250g is perfect for routine laboratory work where precise pH adjustments and accurate reagent quantities are essential for reproducible outcomes.
Catalog No. 50-006-61302 Supplier Alkali Scientific Supplier No. POA005CP
Potassium Phosphate Monobasic, ACS Grade, in a 250g volume is a versatile laboratory reagent used to prepare buffer solutions for a wide range of biochemical, microbiological, and chemical applications. It is especially useful in preparing phosphate buffers and phosphate-buffered saline (PBS) for enzymatic reactions, protein assays, and nucleic acid extractions. Potassium Phosphate Monobasic helps maintain the desired pH in various laboratory processes, ensuring consistency and reliability in experimental results. This smaller volume of 250g is perfect for routine laboratory work where precise pH adjustments and accurate reagent quantities are essential for reproducible outcomes.