Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Monobasic, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18605672 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-672 125 g Poly Bottle
18-605-673 500 g Poly Bottle
18-605-675 2.5 kg Poly Bottle
18-605-674 6 x 500 g Poly Bottle
18-605-676 4 x 2.5 kg Poly Bottle
18-605-677 12 kg Poly Pail
18-605-678 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-605-672 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1380125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

P1380, 7778-77-0, KH2PO4

Spectrum™ Chemical Potassium Phosphate Monobasic, Crystal, Reagent, ACS is a soluble salt that is commonly used as a buffering agent. As an ACS grade Reagent, Spectrum Chemical manufactured potassium phosphate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

pH 4.1 to 4.5
CAS Max % 1
Quantity 125 g
Percent Purity 99%
Grade ACS
Loss on Drying 0.002
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.