Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium phosphate monobasic, meets analytical spec. of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance), Solstice

Catalog No. 60044540 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 500 g
4 x 5 kg
6 x 1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-540 6 x 1 kg
60-018-27 1 kg
60-018-29 5 kg
60-044-539 4 x 5 kg
60-018-28 500 g
60-044-541 6 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-044-540 Supplier Solstice Supplier No. 042436X1KG
Add to Cart
Add to Cart

Meets analytical specification of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

pH 4.2 - 4.5
CAS Max % 100.0000%
Quantity 6 x 1 kg
Assay Percent Range 98 to 100.5%
Linear Formula KH2PO4
IUPAC Name potassium;dihydrogen phosphate
Formula Weight 136.09
Grade Reagent Ph. Eur.
Loss on Drying max. 1.0%
Packaging Poly Bottle
Density 2.34 g/cm3
Chemical Name or Material Potassium phosphate monobasic, Meets analytical specification of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance)
Show More Show Less
ShelfLife 1260 days from date of manufacture
EINECSNumber 231-913-4
RTECSNumber TC6615500
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.