Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Monobasic, MP Biomedicals™

Catalog No. MP021954531 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021954531 100 g
MP021954535 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021954531 Supplier MP Biomedicals, Inc Supplier No. 02195453.1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Displays a high buffering capacity. Potassium Phosphate Monobasic, MP Biomedicals is a reagent that is used as a component for a wide variety of media used in the culture of microorganisms and in phosphate buffered saline (PBS). In addition to helping mai

It occurs in several forms: monobasic, dibasic and tribasic (K3PO4). Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 100 g
Solubility Information Soluble in water; Insoluble to slightly soluble in ethanol; insoluble in alcohol.
Formula Weight 136.09
Chemical Name or Material Potassium Phosphate
Recommended Storage Room Temperature (15–30°C), desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.