Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Monobasic, Supelco

Catalog No. 11100265 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-265 100 g
11-100-253 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-265 Supplier MilliporeSigma Supelco Supplier No. 5438410100
Add to Cart
Add to Cart

For HPLC LiChropur™

  • Unlocking the potential of biochar: Potassium phosphate monobasic is utilized to enhance the adsorption capacity of biochar composites for removing heavy metals like Pb(II) and Cd(II) from water, highlighting its effectiveness in environmental cleanup strategies (Li et al., 2024).
  • Partitioning recombinant chitinase from Nicotiana benthamiana by an aqueous two-phase system: Demonstrates the application of potassium phosphate monobasic in aqueous two-phase systems for efficient partitioning of enzymes, which is crucial for biotechnological and industrial applications (Tue et al., 2024).
  • Unlocking Nature's Toolbox: This study showcases how potassium phosphate monobasic can be used to optimize conditions for the production of recombinant proteins, enhancing yields and streamlining production processes in biomanufacturing (Rajak et al., 2024).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium phosphate monobasic
CAS 7778-77-0
Physical Form Solid
Quantity 100 g
Molecular Weight (g/mol) 136.09
Percent Purity Unknown
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.