Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Phosphate, Tribasic
SCI_ED

Catalog No. s25501 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25501 100 g Reagent Grade
S25501A 500 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25501
Only null left
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7778-53-2
Molecular Formula K3O4P
Molecular Weight (g/mol) 212.265
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
Synonym potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321
PubChem CID 62657
IUPAC Name tripotassium;phosphate
SMILES [O-]P(=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Color White
Quantity 100 g
Assay Percent Range >95%
Linear Formula K3PO4·nH2O
Formula Weight 212.03
Packaging Reagent Grade
Chemical Name or Material Potassium Phosphate, Tribasic
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 8, II, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.