Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Phosphate Tribasic Monohydrate

Catalog No. 04249500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-249-500G 500 g
04-249-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 04-249-500G Supplier Sigma Aldrich Supplier No. 04249500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium phosphate tribasic can be used in Suzuki-Miyaura cross-coupling reactions of +a-boronated esters with aromatic halides to synthesize regioselectively substituted amino esters with the retention of configurations.

Specifications

CAS 27176-10-9
Quantity 500 g
Molecular Formula K3O4P -+ H2O
Linear Formula K3O4P -+ H2O
MDL Number MFCD00150410
Synonym tert-Potassium phosphate monohydrate; Tripotassium phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.