Learn More
Apexbio Technology LLC Potassium phosphate monobasic 7778-77-0 500g
SDP
Catalog No. 502240752
Encompass_Preferred
Potassium phosphate monobasic (7778-77-0) is a phosphate and potassium ion source widely used as a buffering reagent in biochemical and molecular biology applications It acts to maintain stable pH conditions in biological reaction systems thereby supporting enzyme activities and cellular processes Based on these properties potassium phosphate monobasic holds research potential in the preparation of physiological buffers tissue culture media analytical reagents and nutrient mixtures within biological research settings
This item is not returnable. View return policy
Catalog No. 50-224-0752 Supplier Apexbio Technology LLC Supplier No. B7291500000
Potassium phosphate monobasic (7778-77-0) is a phosphate and potassium ion source widely used as a buffering reagent in biochemical and molecular biology applications It acts to maintain stable pH conditions in biological reaction systems thereby supporting enzyme activities and cellular processes Based on these properties potassium phosphate monobasic holds research potential in the preparation of physiological buffers tissue culture media analytical reagents and nutrient mixtures within biological research settings