Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium pyrophosphate

Catalog No. 32243125KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-243-125KG 2.5 kg
32-243-1500G 500 g
32-243-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 32-243-125KG Supplier Sigma Aldrich Supplier No. 3224312.5KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium pyrophosphate

Specifications

Melting Point 1300°C
CAS 7320-34-5
Quantity 2.5 kg
Molecular Formula K4O7P2
Linear Formula K4P2O7
MDL Number MFCD00011393
Synonym Potassium diphosphate tetrabasic; Tetrapotassium diphosphate
Molecular Weight (g/mol) 330.34
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.