Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium silver cyanide, 99.9% (metals basis), Ag 54.2%

Catalog No. AA1255122
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1255122 100 g
AA1255114 25 g
AA1255136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1255122 Supplier Thermo Scientific Chemicals Supplier No. 01255122
Only null left
Add to Cart

Silver plating | CAS: 506-61-6

Silver platingPotassium silver cyanide is used in silver plating and in the manufacturing of antiseptics. It is a versatile bridging ligand and finds use for multidimensional polymer construction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-61-6
MDL Number MFCD00036297
Synonym Potassium dicyanoargentate; Silver(I) potassium cyanide

Specifications

Color White
Physical Form Powder
Quantity 100 g
Linear Formula KAg(CN)2
UN Number UN1588
Merck Index 14,7669
Solubility Information Soluble in waterSoluble in water; Insoluble in acids.
Formula Weight 199.01
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Ag 54.2%
Sensitivity Light sensitive
Density 2.36 g/mL
Assay (metals basis)
Chemical Name or Material Potassium silver cyanide
Show More Show Less
Hazard Category H300+H310+H330
Hazard Statement GHS H Statement
H300-H310-H330-H318-H290-H315
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
May be corrosive to metals.
Causes skin irritation.
Precautionary Statement P261-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.
EINECSNumber 208-047-0
RTECSNumber TT5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.