Alfa Aesar™ Potassium silver cyanide, 99.9% (metals basis), Ag 54.2%

Silver plating

Manufacturer: Alfa Aesar™ 1255136

Quantity 500g

 View more versions of this product

Catalog No. AA1255136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 506-61-6
    Linear Formula KAg(CN)2
    Formula Weight 199.01
    MDL Number MFCD00036297
    UN Number UN1588
    Merck Index 14,7669
    Synonym Potassium dicyanoargentate; Silver(I) potassium cyanide
    Chemical Name or Material Potassium silver cyanide

    Specifications

    Formula Weight 199.01
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color White
    RTECS Number TT5775000
    TSCA TSCA
    MDL Number MFCD00036297
    UN Number UN1588
    Synonym Potassium dicyanoargentate; Silver(I) potassium cyanide
    Quantity 500g
    Density 2.36g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.
    Merck Index 14,7669
    Solubility Information Soluble in water
    Health Hazard 1 Danger
    Concentration Ag 54.2%
    Physical Form Powder
    Sensitivity Light Sensitive
    Linear Formula KAg(CN)2
    CAS 506-61-6
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H318-H290-H315
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes serious eye damage.
    May be corrosive to metals.
    Causes skin irritation.
    Chemical Name or Material Potassium silver cyanide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H300-H310-H330-H318-H290-H315
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes serious eye damage.
    May be corrosive to metals.
    Causes skin irritation.

    GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit