Learn More
Strem, An Ascensus Company CAS# 11135-81-2. 5g. Potassium sodium alloy 78:22 (99.95%). MFCD00134088
SDP
Catalog No. 5090110605
Encompass
CAS# 11135-81-2. 5g. Potassium sodium alloy 78:22 (99.95%). MFCD00134088. Molecular Weight: . Molecular Formula: NaK. Color/form: liq.; 78% K, 22% Na (under argon) Strem# 19-1910. http://www.strem.com/catalog/v/19-1910/
Catalog No. 50-901-10605 Supplier Strem, An Ascensus Company Supplier No. 1919105G
CAS# 11135-81-2. 5g. Potassium sodium alloy 78:22 (99.95%). MFCD00134088. Molecular Weight: . Molecular Formula: NaK. Color/form: liq.; 78% K, 22% Na (under argon) Strem# 19-1910. http://www.strem.com/catalog/v/19-1910/