Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Sodium Tartrate Tetrahydrate

Catalog No. S23771KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S23771KG 1 kg
S23775KG 5 kg
S2377500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S23771KG Supplier Sigma Aldrich Supplier No. S23771KG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium sodium tartrate tetrahydrate is a ferroelectric crystal with a high piezoelectric effect and electromechanical coupling coefficient.Potassium sodium tartrate tetrahydrate has been used in the preparation of 3,5-dinitrosalicylic acid solution used in the determination of the reducing sugar.ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 6106-21-4
Molecular Formula C4H4Na2O4 -+ 6H2O
Linear Formula NaOOCCH2CH2COONa -+ 6H2O
Quantity 1 kg
MDL Number MFCD00149117
Synonym L(+)-Tartaric acid potassium Sodium salt; Rochelle salt; Seignette salt
Molecular Weight (g/mol) 270.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.