Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Sodium Tartrate Tetrahydrate

Catalog No. 323121KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-312-1KG 1 kg
32-312-500G 500 g
32-312-250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 32-312-1KG Supplier Sigma Aldrich Supplier No. 323121KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium Sodium Tartrate Tetrahydrate

Specifications

CAS 6381-59-5
Molecular Formula C4H4KNaO6 -+ 4H2O
Linear Formula KOCOCH(OH)CH(OH)COONa -+ 4H2O
MDL Number MFCD00150989
Quantity 1 kg
Synonym L(+)-Tartaric acid potassium Sodium salt; Rochelle salt; Seignette salt
Molecular Weight (g/mol) 282.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.