Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sodium Tartrate Tetrahydrate, ACS Reagent (Rochelle Salt), Reagents

Catalog No. 01337789
Change view
Click to view available options
Quantity:
100 mL
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-789 500 g
01-337-792 100 mL
01-337-790 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-789 Supplier Reagents Holdings Llc Supplier No. C1290500500A5
Add to Cart
Add to Cart

Potassium Sodium Tartrate Tetrahydrate, ACS Reagent (Rochelle Salt), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.22
InChI Key VZOPRCCTKLAGPN-ZFJVMAEJSA-L
Synonym Rochelle Salt, Sodium potassium tartrate tetrahydrate
IUPAC Name potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
SMILES O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Boiling Point 220°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.79
Chemical Name or Material Potassium Sodium Tartrate Tetrahydrate, ACS Reagent (Rochelle Salt)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.