Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sodium Tartrate Tetrahydrate, EMSURE™, ACS, MilliporeSigma™

Catalog No. M1080870500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1080870500 500 g Plastic Bottle
M1080871000 1 kg Plastic Bottle
M1080875000 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1080870500 Supplier MilliporeSigma Supplier No. 1.08087.0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.218
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]

Specifications

Melting Point 70°C to 80°C
Color White
pH 7.0 to 8.5 (50g/L, H2O, 25°C)
Physical Form Solid
Quantity 500 g
Solubility Information 630g/L
Formula Weight 282.23
Percent Purity 99.0 to 102.0%
Grade ACS/ISO/Reagent Ph. Eur.
Packaging Plastic Bottle
EINECSNumber 206-156-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.