Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sodium Tartrate, Tetrahydrate, Granular, USP, 99-102%, Spectrum™ Chemical
SDP

Catalog No. 18605702 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-702 50 kg Fiber Drum
18-605-699 500 g Poly Bottle
18-605-700 2.5 kg Poly Pail
18-605-701 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-702 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO20150KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PO201, 6381-59-5, C4H4KNaO6·4H2O

Spectrum™ Chemical Potassium Sodium Tartrate, Tetrahydrate, Granular, USP is sometimes referred to as Rochelle or Seignette's salt and used as a laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.22
InChI Key VZOPRCCTKLAGPN-ZFJVMAEJSA-L
IUPAC Name potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
SMILES O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

CAS Max % 1
Packaging Fiber Drum
Quantity 50 kg
Percent Purity 99 to 102%
Grade USP
Physical Form Crystalline Powder
Chemical Name or Material Potassium Sodium Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.