Learn More
Potassium Sodium Tartrate Tetrahydrate, Purified (Rochelle Salt), Reagents
Specifications
Specifications
| Chemical Name or Material | Potassium Sodium Tartrate Tetrahydrate, Purified (Rochelle Salt) |
| Boiling Point | 220°C |
| CAS | 6381-59-5 |
| Color | Colorless/White |
| Physical Form | Solid |
| Quantity | 500 g |
| Packaging | Poly Bottle |
| Specific Gravity | 1.79 |
| Molecular Formula | KNaC4H4O6, 4H2O |
| Synonym | Rochelle Salt, Sodium potassium tartrate tetrahydrate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.