Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sodium Tartrate Tetrahydrate, Purified (Rochelle Salt), Reagents

Catalog No. 01337090
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-090 3 kg
01-337-089 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-090 Supplier Reagents Holdings Llc Supplier No. C22905003A5
Add to Cart
Add to Cart

Potassium Sodium Tartrate Tetrahydrate, Purified (Rochelle Salt), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Sodium Tartrate Tetrahydrate, Purified (Rochelle Salt)
Boiling Point 220°C
CAS 6381-59-5
Color Colorless/White
Physical Form Solid
Quantity 3 kg
Molecular Formula KNaC4H4O6, 4H2O
Synonym Rochelle Salt, Sodium potassium tartrate tetrahydrate
Molecular Weight (g/mol) 282.2201
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.79
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.