Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium, solid, 99% (metals basis)

Catalog No. aa4584203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4584203 1 g
AA4584206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4584203 Supplier Thermo Scientific Chemicals Supplier No. 04584203
Only null left
Add to Cart

CAS: 7440-09-7 | K | 39.10 g/mol

Potassium combines under pressure with silver metal to form 2 new compounds: K2Ag and K3Ag. This chemical is important in neuron (brain and nerve) function, and in influencing osmotic balance between cells and the interstitial fluid, with their distribution mediated in all animals (but not in all plants) by the so-called Na+/K+-ATPase pump. It is also used to prevent muscle contraction and the sending of all nerve impulses in animals through action potentials. Potassium ion is a nutrient necessary for human life and health. It is used as reducing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-09-7
Molecular Formula K
Molecular Weight (g/mol) 39.10
MDL Number MFCD00133776
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
Synonym kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
PubChem CID 5462222
ChEBI CHEBI:26216
IUPAC Name potassium
SMILES [K]

Specifications

Quantity 1 g
Odor Odorless
Packaging Breakseal Ampules
UN Number UN2257
Solubility Information Soluble in water.
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Potassium
Hazard Category H260-H314-H335
Hazard Statement GHS H Statement
H260-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: POTASSIUM
EINECSNumber 231-119-8
RTECSNumber TS6460000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.