Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Sorbate

Catalog No. 85520250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-520-250G 50 g
85-520-1KG 250 g
85-520-50G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 85-520-250G Supplier Sigma Aldrich Supplier No. 85520250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium sorbate is potassium salt of sorbic acid. Qualitative and quantitative estimation of potassium sorbate in foods and beverages-ßby high-performance liquid chromatography (HPLC) followed by UV diode array detection (HPLC-UV)-ßand LC-MS/MS (Liquid chromatography-mass spectrometry)-ßhave been proposed.

Specifications

CAS 24634-61-5
Molecular Formula C6H7KO2
Linear Formula CH3CH=CHCH=CHCOOK
MDL Number MFCD00016546
Quantity 50 g
Synonym Potassium 2,4-hexadienoate; Sorbic acid potassium salt
Molecular Weight (g/mol) 150.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.