Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Sorbate

Catalog No. 8552050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-520-50G 1 g
85-520-1KG 250 g
85-520-250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 85-520-50G Supplier Sigma Aldrich Supplier No. 8552050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium sorbate is potassium salt of sorbic acid. Qualitative and quantitative estimation of potassium sorbate in foods and beverages-ßby high-performance liquid chromatography (HPLC) followed by UV diode array detection (HPLC-UV)-ßand LC-MS/MS (Liquid chromatography-mass spectrometry)-ßhave been proposed.

Specifications

CAS 1002-57-9
Melting Point 194°C
Molecular Formula C8H17NO2
Linear Formula NH2(CH2)7COOH
MDL Number MFCD00008245
Quantity 1 g
Synonym Potassium 2,4-hexadienoate; Sorbic acid potassium salt
Molecular Weight (g/mol) 159.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.