Learn More
Medchemexpress LLC Potassium sorbate | 24634-61-5 | 150.22 | 500 G
SDP
Catalog No. 5000388638
Encompass_Preferred
Potassium sorbate is an antibacterial preservative that effectively inhibits the growth of most molds, yeasts, and various bacterial species. It has been shown to suppress the activity of intestinal microorganisms and alter intestinal flora composition in model organisms, while also reducing levels of intestinal immune biomarkers such as IgG, IL-1β, and TNF-α. This compound is valuable in research for its potential in preventing and treating mold, yeast, and bacterial infections.
  • Effective antibacterial preservative against molds, yeasts, and bacteria.
  • Inhibits microbial growth.
  • Suppresses intestinal microorganism activity.
  • Alters intestinal flora composition.
  • Reduces levels of intestinal immune biomarkers.
  • Useful in research for infection prevention and treatment.
Catalog No. 50-003-88638 Supplier Medchemexpress LLC Supplier No. HYN0626A500G
Potassium sorbate is an antibacterial preservative that effectively inhibits the growth of most molds, yeasts, and various bacterial species. It has been shown to suppress the activity of intestinal microorganisms and alter intestinal flora composition in model organisms, while also reducing levels of intestinal immune biomarkers such as IgG, IL-1β, and TNF-α. This compound is valuable in research for its potential in preventing and treating mold, yeast, and bacterial infections.
  • Effective antibacterial preservative against molds, yeasts, and bacteria.
  • Inhibits microbial growth.
  • Suppresses intestinal microorganism activity.
  • Alters intestinal flora composition.
  • Reduces levels of intestinal immune biomarkers.
  • Useful in research for infection prevention and treatment.