Learn More
Potassium Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™
Shop All Spex CertiPrep Products

Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Potassium Standard
- Made with ASTM Type I Water
- 1,000 μg/mL concentration
- 250 mL volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Specifications
Specifications
| Packaging | Poly Bottle |
| Certificate of Analysis | Assurance Certificate of Analysis reports up to 30 elements at ppb levels. |
| Technique | AA, ICP |
| Type | Certified Reference Material |
| Quantity | 250 mL |
| Grade | Assurance |
| Chemical Name or Material | Potassium Standard |
Safety and Handling
| Hazard Category | Causes skin irritation, serious eye irritation |
| DOTInformation | Nonhazardous |
| Recommended Storage | Ambient |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.