Learn More
Medchemexpress LLC Potassium stearate | 593-29-3 | MFCD00072385 | 98.0% | 322.57 g/mol | C18H35KO2 | 100 G
SDP
Catalog No. 5000283408
Encompass_Preferred
Potassium stearate is the potassium salt of stearic acid supplied as a 98% pure biochemical assay reagent. It has molecular formula C18H35KO2 and a molecular weight of 322.57 g/mol. The material is commonly used as a surfactant, emulsifier, and reagent in laboratory assays and formulations and is supplied as a solid for research use.
  • High purity (98%) for analytical and research applications.
  • Acts as a surfactant and emulsifier in formulations.
  • Suitable for use as a reagent in biochemical assays.
  • Solid form for easy handling and storage.
  • Datasheet provides handling and safety information for laboratory use.
Catalog No. 50-002-83408 Supplier Medchemexpress LLC Supplier No. HYW133986100G
Potassium stearate is the potassium salt of stearic acid supplied as a 98% pure biochemical assay reagent. It has molecular formula C18H35KO2 and a molecular weight of 322.57 g/mol. The material is commonly used as a surfactant, emulsifier, and reagent in laboratory assays and formulations and is supplied as a solid for research use.
  • High purity (98%) for analytical and research applications.
  • Acts as a surfactant and emulsifier in formulations.
  • Suitable for use as a reagent in biochemical assays.
  • Solid form for easy handling and storage.
  • Datasheet provides handling and safety information for laboratory use.