Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Potassium sulfate, 99+%, for biochemistry

Catalog No. AC424212500
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424212500 250 g Plastic Bottle
AC424210010 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC424212500 Supplier Thermo Scientific Chemicals Supplier No. 424212500
  • $84.00
    $76.65 / Each of 1

    Save $7.35

    9% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7778-80-5 | K2O4S | 174.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689.0°C
Melting Point 1067.0°C
Color White
Physical Form Crystals
Quantity 250 g
Packaging Plastic Bottle
Assay Percent Range 99+%
Linear Formula K2SO4
Merck Index 15,7793
Solubility Information (10 g in 150 ml water) Clear colorless
Formula Weight 174.25
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material Potassium sulfate
Show More Show Less
EINECSNumber 231-915-5
RTECSNumber TT5900000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.