Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sulfate, Crystal, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18612910 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-910 50 kg Poly Bottle
18-612-906 125 g Poly Bottle
18-612-907 500 g Poly Bottle
18-612-908 2.5 kg Poly Bottle
18-612-909 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-612-910 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P140550KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

pH 5.5 to 8.5
CAS Max % 1
Physical Form Crystals or Needles
Quantity 50 kg
Percent Purity 99%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Sulfate, Crystal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.