Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sulfate, FCC, 99-100.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7778-80-5
K2O4S
174.25
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium sulfate
[K+].[K+].[O-]S([O-])(=O)=O
7778-80-5
K2O4S
174.25
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium sulfate
[K+].[K+].[O-]S([O-])(=O)=O

P1589, 7778-80-5, K2SO4

Spectrum™ Potassium Sulfate, FCC is used as an acidity regulator. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Percent Purity 99 to 100.5%
Grade FCC
Lead (Pb) 2 mg/kg
Packaging Poly Bottle
Selenium (Se) 5 mg/kg
Quantity 125 g