Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium sulfate, meets analytical spec. of Ph. Eur., 99-101%, Solstice

Catalog No. 60044747 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-747 6 x 1 kg Poly Bottle
60-019-54 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-747 Supplier Solstice Supplier No. 126586X1KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Meets analytical specification of Ph. Eur., 99-101%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1,698°C
CAS Max % 100.0000%
Physical Form Powder
Quantity 6 x 1 kg
Assay Percent Range 99 to 101%
Linear Formula K2SO4
UN Number NONH for all modes of transport
Solubility Information Insoluble in alcohol
IUPAC Name dipotassium sulfate
Formula Weight 174.26
Percent Purity 99 to 101%
Grade Reagent Ph. Eur.
Packaging Poly Bottle
Density 2.66 g/cm3
Chemical Name or Material Potassium sulfate, Meets analytical specification of Ph. Eur., 99-101%
Show More Show Less
ShelfLife 1260 days from date of manufacture
EINECSNumber 231-915-5
RTECSNumber TT5900000
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.