Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sulfate, (Powder/ACS Reagent Grade), MP Biomedicals

Catalog No. ICN15256491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15256491 1 kg
ICN15256490 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15256491 Supplier MP Biomedicals, Inc Supplier No. 0215256491
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 1 kg
IUPAC Name dipotassium sulfate
Formula Weight 174.26
Grade ACS Reagent
Chemical Name or Material Potassium Sulfate, Anhydrous
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.