Learn More
Hach Company Potassium sulfate sol. Sat.
Catalog No. 5000207033 Shop All Hach Company Products
Encompass
KS160 Potassium sulfate Solution-Saturated-500 mL (Radiometer Analytical) Filling and storage solution for combined/reference electrodes 500 mL
Catalog No. 50-002-07033 Supplier Hach Company Supplier No. C20C500
KS160 Potassium sulfate Solution-Saturated-500 mL (Radiometer Analytical) Filling and storage solution for combined/reference electrodes 500 mL