Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Sulfite

Catalog No. 6585103KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 kg
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-851-03KG 3 kg
65-851-025G 25 g
65-851-01KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 65-851-03KG Supplier Sigma Aldrich Supplier No. 6585103KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium sulfite (K2SO3) can be used as a catalyst in the oxidation of sulfur dioxide. It is also used as an anionic catalyst for the cyclotrimerization of aryl isocyanates to prepare heterocyclic symmetrical isocyanurates.

Specifications

CAS 10117-38-1
Quantity 3 kg
Molecular Formula K2O3S
Linear Formula K2SO3
MDL Number MFCD00011387
Molecular Weight (g/mol) 158.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.