Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sulphite, MP Biomedicals

Catalog No. ICN225855 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225855 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225855 Supplier MP Biomedicals, Inc Supplier No. 0522585525
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10117-38-1
Molecular Formula K2O3S
Molecular Weight (g/mol) 158.254
InChI Key BHZRJJOHZFYXTO-UHFFFAOYSA-L
Synonym potassium sulfite, dipotassium sulfite, potassium sulphite, sulfurous acid, dipotassium salt, unii-015kzc652e, sulfurous acid, potassium salt 1:2, potassium sulfite k2so3, kaliumsulfit, potassiumsulfite, dipotassium ion sulfite
PubChem CID 24958
IUPAC Name dipotassium;sulfite
SMILES [O-]S(=O)[O-].[K+].[K+]

Specifications

Quantity 25 g
Formula Weight 158.26
Chemical Name or Material Potassium Sulphite
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.