Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Tartrate Dibasic Hemihydrate

Catalog No. 25510500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-510-500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 25-510-500G Supplier Sigma Aldrich Supplier No. 25510500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium tartrate dibasic hemihydrate is the dipotassium salt of tartaric acid.Potassium tartrate dibasic hemihydrate (Potassiu mL-tartrate dibasic hemihydrate) may be used in the hydrothermal synthesis of 4 metal-organic coordination polymers:{[Cd3(C4H4O6)3(H2O)].H2O}n {Cd3(C4H3O6)2(H2O)2].5.5H2O}n {[Cd2(C4H4O6)2(H2O)].3H2O}n[Cd(C4H4O6)]n

Specifications

CAS 563-43-9
Melting Point 32°C
Density 1.207 g/mL at 25°C
Boiling Point 115°C (50 mmHg, lit.)
Molecular Formula C2H5AlCl2
Linear Formula C2H5AlCl2
Quantity 500 g
MDL Number MFCD00000457
Synonym L-(+)-Tartaric acid dipotassium salt; Dipotassium tartrate hemihydrate
Molecular Weight (g/mol) 126.95 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.