Learn More
MOLTOX Molecular Toxicology Inc Potassium Tellurite Agar, 20 mL/plate, 20 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948700
Encompass_Preferred
Potassium Tellurite Agar is a selective medium used for the isolation and cultivation of Corynebacterium. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-700 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S91
Potassium Tellurite Agar is a selective medium used for the isolation and cultivation of Corynebacterium. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.