Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tellurite monohydrate, 97%

Catalog No. AA4195914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4195914 25 g
AA4195922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4195914 Supplier Thermo Scientific Chemicals Supplier No. 04195914
Add to Cart

CAS: 123333-66-4 | K2O3Te·H2O

Potassium tellurite, in association with agar is used as a selective medium for growth of clauberg (bacteria). It is used for the selective isolation of staphylococci and corynebacteria, which is used to reduce TeO3 2- to tellurium element.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-66-4
Molecular Formula K2O3Te·H2O
MDL Number MFCD00149923
Synonym potassium tellurite, potassium tellurate iv, dipotassium tellurite, dipotassium trioxotellurate, telluric acid, dipotassium salt, potassiium tellurite iv, telluric acid h2teo3 , dipotassium salt, dipotassium ion tellurite, acmc-1bm2e

Specifications

Melting Point 465°C (decomposition)
Color White
Physical Form Crystals
Quantity 25 g
Assay Percent Range 97%
Solubility Information Soluble in water.
Formula Weight 253.80 Anhydrous
Appearance White crystals
Chemical Name or Material Potassium tellurite monohydrate
Hazard Category H301
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation DOT Class: 6.1; Packaging Group: III
EINECSNumber 232-213-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.