Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium tert-butoxide solution

Catalog No. 32865050ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
500 mL
2 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-865-050ML 50 mL
32-865-020L 20 L
32-865-02L 2 L
32-865-0500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 32-865-050ML Supplier Sigma Aldrich Supplier No. 32865050ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium tert-butoxide solution is a strong non nucleophilic base. It is capable of deprotonating carbon and other Bronsted acids. Reagent solutions in tert-BuOh may be prepared by reacting anhydrous alcohol with potassium in nitrogen atmosphere.Potassium tert-butoxide is used to prepare 3-potassiooxamethylpyridine catalyst. It is also used as a strong alkoxide base reagent.

Specifications

CAS 865-47-4
Density 0.902 g/mL (at 25°C)
Molecular Formula C4H9KO
Linear Formula (CH3)3 COK
Quantity 50 mL
MDL Number MFCD00012162
Synonym Potassium tert-butylate; Potassium-2-methylpropan-2-olate solution
Molecular Weight (g/mol) 112.21
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.