Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tert-pentoxide, 0.9M (14 wt.%) solution in cyclohexane, AcroSeal™

Catalog No. AC164721000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164721000 100 mL AcroSeal™ Glass bottle
AC164728000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC164721000 Supplier Thermo Scientific Chemicals Supplier No. 164721000
Add to Cart

CAS: 41233-93-6 | C5H11KO | 126.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium tert-pentoxide
CAS 41233-93-6, 110-82-7
Quantity 100 mL
Molecular Formula C5H11KO
Linear Formula C2H5C(CH3)2OK
MDL Number MFCD00064808
Fieser 01,939
Flash Point −18°C
Synonym potassium 2-methylbutan-2-olate, potassium tert-amylate, potassium tert-pentoxide, potassium tert-pentylate, potassium tert-pentoxide solution, potassium 2-methyl-2-butoxide, 2-butanol, 2-methyl-, potassium salt, potassium tert-amyloxide, 2-butanol, 2-methyl-, potassium salt 1:1
Solubility Information Solubility in water: hydrolyzes
InChI Key ZRLVQFQTCMUIRM-UHFFFAOYSA-N
SMILES [K+].CCC(C)(C)[O-]
IUPAC Name potassium 2-methylbutan-2-olate
Molecular Weight (g/mol) 126.24
PubChem CID 23683543
Formula Weight 126.24
Packaging AcroSeal™ Glass bottle
Refractive Index 1.4211
Specific Gravity 0.82
Density 0.8200g/mL
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.