Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tert-Pentoxide (ca. 15% in Cyclohexane, ca. 1mol/L), TCI America™
SDP

Catalog No. P224625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P224625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P224625ML Supplier TCI America Supplier No. P224625ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41233-93-6
Molecular Formula C5H11KO
Molecular Weight (g/mol) 126.24
MDL Number MFCD00064808
InChI Key ZRLVQFQTCMUIRM-UHFFFAOYSA-N
Synonym potassium 2-methylbutan-2-olate, potassium tert-amylate, potassium tert-pentoxide, potassium tert-pentylate, potassium tert-pentoxide solution, potassium 2-methyl-2-butoxide, 2-butanol, 2-methyl-, potassium salt, potassium tert-amyloxide, 2-butanol, 2-methyl-, potassium salt 1:1
PubChem CID 23683543
IUPAC Name potassium 2-methylbutan-2-olate
SMILES [K+].CCC(C)(C)[O-]

Specifications

Color Yellow
Quantity 25 mL
UN Number 2924
Formula Weight 126.24
Physical Form Liquid
Chemical Name or Material Potassium tert-Pentoxide (ca. 15% in Cyclohexane, ca. 1mol/L)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.