Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Tert-Pentoxide Solution

Catalog No. 60435250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-435-250ML 250 mL
60-435-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 60-435-250ML Supplier Sigma Aldrich Supplier No. 60435250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium tert-pentoxide solution has been used as a base to synthesize [RuClPh(CO)(H2IMes)(PCy3)] from second-generation Grubbs metathesis catalyst, [RuCl2(=CHPh)(H2IMes)(PCy3)], (where H2IMes is 1,3-dimethyl-4,5-dihydroimidazol-2-ylidene and PCy3 is tricyclohexylphosphine) and benzyl alcohol. This complex is a solid, air-stable and highly active isomerization catalyst.

Specifications

CAS 41233-93-6
Density 0.90 g/mL at 20°C
Molecular Formula C5H11KO
Linear Formula CH3CH2C(CH3)2OK
MDL Number MFCD00064808
Quantity 250 mL
Synonym Potassium (2-methyl-2-butoxide); Potassium tert-pentylate
Molecular Weight (g/mol) 126.24 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.