Learn More
Strem, An Ascensus Company CAS# 10025-98-6. 1g. Potassium tetrachloropalladate(II), 99%. MFCD00011373
SDP
Supplier:  Strem, An Ascensus Company 4621261G
Encompass
CAS# 10025-98-6. 1g. Potassium tetrachloropalladate(II), 99%. MFCD00011373. Molecular Weight: 326.41. Molecular Formula: K2PdCl4. Color/form: brown pwdr. Strem# 46-2126. http://www.strem.com/catalog/v/46-2126/
Catalog No. 50-901-12469