Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Tetracyanoborate 98.0+%, TCI America™
SDP

Catalog No. P2168200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2168200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2168200MG Supplier TCI America Supplier No. P2168200MG
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 261356-49-4
Molecular Formula C4BKN4
Molecular Weight (g/mol) 153.98
MDL Number MFCD11983240
InChI Key SQFZXQOCCKUPEC-UHFFFAOYSA-N
PubChem CID 23686396
IUPAC Name potassium;tetracyanoboranuide
SMILES [B-](C#N)(C#N)(C#N)C#N.[K+]

Specifications

Melting Point 413°C
Color White-Yellow
Quantity 200 mg
UN Number 3439
Formula Weight 153.98
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Tetracyanoborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.